Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(1-Piperazinyl)acetophenone, 94%

Chemical Identifiers

Chemical Name or Material 4'-(1-Piperazinyl)acetophenone
CAS 51639-48-6
Molecular Formula C12H16N2O
Formula Weight 204.27
MDL Number MFCD00005962
UN Number UN3259
Synonym 4'-(1-Piperazino)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2283206

 
alfa aesar™
B2283206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2283214

 
alfa aesar™
B2283214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2283222

 
alfa aesar™
B2283222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.27
Molecular Formula C12H16N2O
MDL Number MFCD00005962
CAS 51639-48-6
UN Number UN3259
Melting Point 93° to 98°C
Quantity 5g
Chemical Name or Material 4'-(1-Piperazinyl)acetophenone
Assay 94%
Synonym 4'-(1-Piperazino)acetophenone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.