Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Oxo-5Z,8Z,11Z,14Z-eicosatetraenylamino)butanoic acid, 98%

Chemical Identifiers

Chemical Name or Material 4-(1-Oxo-5Z,8Z,11Z,14Z-eicosatetraenylamino)butanoic acid
CAS 128201-89-8
Formula Weight 389.58
MDL Number MFCD05863981
Synonym N-Arachidonyl-GABA; NAGABA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67423LB0

 
alfa aesar™
J67423LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67423MA

 
alfa aesar™
J67423MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 389.58
Assay 98%
MDL Number MFCD05863981
CAS 128201-89-8
Synonym N-Arachidonyl-GABA; NAGABA
Physical Form Liquid
Color Yellow
Storage Note 1 -30° to -10°C
Quantity 5mg
Sensitivity Air Sensitive
Chemical Name or Material 4-(1-Oxo-5Z,8Z,11Z,14Z-eicosatetraenylamino)butanoic acid
Safety and Handling

-30° to -10°C