Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(1-Imidazolyl)acetophenone, 97%

$87.88 - $340.18

Chemical Identifiers

Chemical Name or Material 4'-(1-Imidazolyl)acetophenone
CAS 10041-06-2
Molecular Formula C11H10N2O
Formula Weight 186.21
MDL Number MFCD00005282
Beilstein 608955
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0611506

 
alfa aesar™
L0611506
5g Each for $87.88

AAL0611514

 
alfa aesar™
L0611514
25g Each for $340.18
Description & Specifications

Specifications

Formula Weight 186.21
Chemical Name or Material 4'-(1-Imidazolyl)acetophenone
Assay 97%
Molecular Formula C11H10N2O
MDL Number MFCD00005282
CAS 10041-06-2
Beilstein 608955
Melting Point 114° to 118°C
Quantity 5g