Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Cyclopenten-1-yl)morpholine, 96%

$56.04 - $173.93

Chemical Identifiers

Chemical Name or Material 4-(1-Cyclopenten-1-yl)morpholine
CAS 936-52-7
Molecular Formula C9H15NO
Formula Weight 153.23
MDL Number MFCD00006162
Beilstein 117154
Synonym 1-(4-Morpholino)cyclopentene; 1-(4-Morpholinyl)cyclopentene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0026214

 
alfa aesar™
L0026214
25g Each for $56.04

AAL0026222

 
alfa aesar™
L0026222
100g Each for $173.93
Description & Specifications

Specifications

Formula Weight 153.23
Chemical Name or Material 4-(1-Cyclopenten-1-yl)morpholine
TSCA TSCA
Assay 96%
Molecular Formula C9H15NO
MDL Number MFCD00006162
CAS 936-52-7
Beilstein 117154
Synonym 1-(4-Morpholino)cyclopentene; 1-(4-Morpholinyl)cyclopentene
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number QE0685000
Flash Point 64°C (147°F)
Boiling Point 105° to 106°C (12mmHg)
Health Hazard 1 Warning
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C