Learn More
4-(1-Bromoethyl)benzoic acid, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific L1302506
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
4-(1-Bromoethyl)benzoic acid | |
155°C to 156°C | |
MFCD00020378 | |
2084039 | |
5g | |
VICCYULHZWEWMB-UHFFFAOYSA-N | |
4-(1-bromoethyl)benzoic acid | |
5146225 | |
98% |
113023-73-7 | |
C9H9BrO2 | |
UN3261 | |
4-1-bromo-ethyl-benzoic acid, 4-1-bromoethyl benzoic acid, 4-1-bromoethyl-benzoic acid, acmc-1c3tf, benzoic acid, 4-1-bromoethyl, benzoic acid,4-1-bromoethyl | |
Insoluble in water. | |
CC(C1=CC=C(C=C1)C(=O)O)Br | |
229.073 | |
229.08 |
Chemical Identifiers
113023-73-7 | |
229.073 | |
VICCYULHZWEWMB-UHFFFAOYSA-N | |
5146225 | |
CC(C1=CC=C(C=C1)C(=O)O)Br |
C9H9BrO2 | |
MFCD00020378 | |
4-1-bromo-ethyl-benzoic acid, 4-1-bromoethyl benzoic acid, 4-1-bromoethyl-benzoic acid, acmc-1c3tf, benzoic acid, 4-1-bromoethyl, benzoic acid,4-1-bromoethyl | |
4-(1-bromoethyl)benzoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only