missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1-Azetidinylsulfonyl)benzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H5308803

 View more versions of this product

Catalog No. AAH5308803



Specifications

Specifications

4-(1-Azetidinylsulfonyl)benzeneboronic acid
1g
C9H12BNO4S
4-azetidin-1-ylsulfonyl phenylboronic acid, 4-azetidin-1-ylsulfonyl phenyl boronic acid, 1-4-boronophenylsulfonyl azetidine, 4-1-azetidinylsulfonyl benzeneboronic acid, 4-1-azetidinylsulfonyl phenylboronic acid, boronic acid,b-4-1-azetidinylsulfonyl phenyl, 4-azetidine-1-sulfonyl phenylboronic acid, acmc-209qgc, 4-azetidine-1-sulfonyl phenyl boronic acid
B(C1=CC=C(C=C1)S(=O)(=O)N2CCC2)(O)O
241.068
241.07
140°C to 144°C
871329-68-9
MFCD07363756
IOGLONDWRZETTA-UHFFFAOYSA-N
[4-(azetidin-1-ylsulfonyl)phenyl]boronic acid
44886919
96%

Chemical Identifiers

871329-68-9
241.068
IOGLONDWRZETTA-UHFFFAOYSA-N
44886919
B(C1=CC=C(C=C1)S(=O)(=O)N2CCC2)(O)O
C9H12BNO4S
MFCD07363756
4-azetidin-1-ylsulfonyl phenylboronic acid, 4-azetidin-1-ylsulfonyl phenyl boronic acid, 1-4-boronophenylsulfonyl azetidine, 4-1-azetidinylsulfonyl benzeneboronic acid, 4-1-azetidinylsulfonyl phenylboronic acid, boronic acid,b-4-1-azetidinylsulfonyl phenyl, 4-azetidine-1-sulfonyl phenylboronic acid, acmc-209qgc, 4-azetidine-1-sulfonyl phenyl boronic acid
[4-(azetidin-1-ylsulfonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents