missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-4-(1-Aminoethyl)phenol, 97%

$81.73 - $1,079.49

Chemical Identifiers

CAS 134855-87-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00143209
InChI Key CDQPLIAKRDYOCB-UHFFFAOYSA-N
Synonym 4-1-aminoethyl phenol, 1-p-hydroxyphenyl ethylamine, phenol, 4-1-aminoethyl, 4-aminoethyl phenol, 1-4-hydroxyphenyl ethylamine, p-oxyphenylathylamin, timtec-bb sbb008453, 4-1-aminoethyl-pheno, 4-1-amino-ethyl-phenol, 1-4-hydroxyphenyl ethanamine
PubChem CID 123594
ChEBI CHEBI:581
IUPAC Name 4-(1-aminoethyl)phenol
SMILES CC(C1=CC=C(C=C1)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6370006 View Documents Alfa Aesar™
H6370006
5g Each for $81.73
 
AAH6370014 View Documents Alfa Aesar™
H6370014
25g Each for $325.39
 
AAH6370022 View Documents Alfa Aesar™
H6370022
100g Each for $1,079.49
 
Description

Description

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

134855-87-1
137.182
CDQPLIAKRDYOCB-UHFFFAOYSA-N
123594
4-(1-aminoethyl)phenol
C8H11NO
MFCD00143209
4-1-aminoethyl phenol, 1-p-hydroxyphenyl ethylamine, phenol, 4-1-aminoethyl, 4-aminoethyl phenol, 1-4-hydroxyphenyl ethylamine, p-oxyphenylathylamin, timtec-bb sbb008453, 4-1-aminoethyl-pheno, 4-1-amino-ethyl-phenol, 1-4-hydroxyphenyl ethanamine
CHEBI:581
CC(C1=CC=C(C=C1)O)N

Specifications

5g
C8H11NO
4-1-aminoethyl phenol, 1-p-hydroxyphenyl ethylamine, phenol, 4-1-aminoethyl, 4-aminoethyl phenol, 1-4-hydroxyphenyl ethylamine, p-oxyphenylathylamin, timtec-bb sbb008453, 4-1-aminoethyl-pheno, 4-1-amino-ethyl-phenol, 1-4-hydroxyphenyl ethanamine
CC(C1=CC=C(C=C1)O)N
137.182
CHEBI:581
97%
134855-87-1
MFCD00143209
CDQPLIAKRDYOCB-UHFFFAOYSA-N
4-(1-aminoethyl)phenol
123594
137.18
(+/-)-4-(1-Aminoethyl)phenol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SJ5950150

Recommended Storage : 2° to 8°C