Please login to your online account to display your discounted pricing

4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC40204DE

 View more versions of this product

Catalog No. CC40204DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 189.24
Quantity 5g
Assay 95%
Molecular Formula C10H7NOS
Assay Percent Range 95%
Packaging Amber Glass Bottle
Chemical Name or Material 4-(1, 3-Thiazol-2-yl)benzaldehyde
CAS 198904-53-9

Specifications

Chemical Name or Material 4-(1, 3-Thiazol-2-yl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H7NOS
Formula Weight 189.24