missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1,3-Oxazol-5-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

$144.34 - $285.83

Chemical Identifiers

CAS 337508-66-4
Molecular Formula C9H6ClNO3S
Molecular Weight (g/mol) 243.66
MDL Number MFCD02681962
InChI Key QCGUTLAPXWBIMM-UHFFFAOYSA-N
Synonym 4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, benzenesulfonyl chloride, 4-5-oxazolyl, chloro 4-1,3-oxazol-5-yl phenyl sulfone
PubChem CID 2776415
IUPAC Name 4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14503CB
SDS View Documents Promotion Details
Fisher Scientific Company
CC14503CB
250mg Amber Glass Bottle
Each for $144.34
 
CC14503DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC14503DA
1g Amber Glass Bottle
Each for $285.83
 
Description

Description

Specifications

Chemical Identifiers

337508-66-4
243.66
QCGUTLAPXWBIMM-UHFFFAOYSA-N
2776415
ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1
C9H6ClNO3S
MFCD02681962
4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, benzenesulfonyl chloride, 4-5-oxazolyl, chloro 4-1,3-oxazol-5-yl phenyl sulfone
4-(1,3-oxazol-5-yl)benzenesulfonyl chloride

Specifications

337508-66-4
100.0
C9H6ClNO3S
Amber Glass Bottle
4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, benzenesulfonyl chloride, 4-5-oxazolyl, chloro 4-1,3-oxazol-5-yl phenyl sulfone
ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1
243.66
243.67
4-(1, 3-Oxazol-5-yl)benzenesulfonyl chloride
96.5
97%
MFCD02681962
250mg
QCGUTLAPXWBIMM-UHFFFAOYSA-N
4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
2776415
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications