Please login to your online account to display your discounted pricing

4-(1,3-Oxazol-5-yl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1, 3-Oxazol-5-yl)aniline
CAS 1008-95-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2O
Formula Weight 160.18
MDL Number MFCD00052192
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DFP00120CB

 
fisher scientific company
DFP00120CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.18
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)aniline
Assay Percent Range 97%
Molecular Formula C9H8N2O
MDL Number MFCD00052192
CAS 1008-95-3
Packaging Amber Glass Bottle
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.