Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1,3-Dioxolan-2-yl)thiobenzamide, 97%

$187.64 - $187.64

Chemical Identifiers

Chemical Name or Material 4-(1,3-Dioxolan-2-yl)thiobenzamide
CAS 175202-43-4
Molecular Formula C10H11NO2S
Formula Weight 209.26g/mol
MDL Number MFCD00085174
Synonym 4-Thiocarbamoylbenzaldehyde ethylene glycol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5185603

 
alfa aesar™
H51856-03
Each for $187.64
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-(1,3-Dioxolan-2-yl)thiobenzamide
CAS 175202-43-4
Assay 97%
Molecular Formula C10H11NO2S
Formula Weight 209.26g/mol
MDL Number MFCD00085174
Synonym 4-Thiocarbamoylbenzaldehyde ethylene glycol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.