Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)benzeneboronic acid pinacol ester

$60.34 - $232.43

Chemical Identifiers

Chemical Name or Material 4-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)benzeneboronic acid pinacol ester
CAS 1218790-48-7
Molecular Formula C19H23BN2O5
Formula Weight 370.21
MDL Number MFCD11113040
Synonym 1,3-Dimethyl-5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene]pyrimidine-2,4,6-trione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5112403

 
alfa aesar™
H5112403
1g Each for $60.34

AAH5112406

 
alfa aesar™
H5112406
5g Each for $232.43
Description & Specifications

Specifications

Formula Weight 370.21
Chemical Name or Material 4-(1,3-Dimethyl-2,4,6-trioxohexahydropyrimidin-5-ylidenemethyl)benzeneboronic acid pinacol ester
Molecular Formula C19H23BN2O5
MDL Number MFCD11113040
CAS 1218790-48-7
Quantity 1g
Synonym 1,3-Dimethyl-5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylidene]pyrimidine-2,4,6-trione