missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one, tech., Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5349903

 View more versions of this product

Catalog No. AAH5349903



Description

Description

Applications
4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4-(1,3,3-Trimethyl-7-oxabicyclo[4.1.0]hept-2-yl)-3-buten-2-one
1.491
190059-33-7
MFCD07784337
ODMUHAHUBCUABS-AATRIKPKSA-N
(E)-4-(4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-5-yl)but-3-en-2-one
11041982
Technical
1.039
1g
C13H20O2
3-buten-2-one,4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-, 3e
CC(=O)C=CC1C(CCC2C1(O2)C)(C)C
208.301
208.3

Chemical Identifiers

190059-33-7
208.301
ODMUHAHUBCUABS-AATRIKPKSA-N
11041982
CC(=O)C=CC1C(CCC2C1(O2)C)(C)C
C13H20O2
MFCD07784337
3-buten-2-one,4-1,3,3-trimethyl-7-oxabicyclo 4.1.0 hept-2-yl-, 3e
(E)-4-(4,4,6-trimethyl-7-oxabicyclo[4.1.0]heptan-5-yl)but-3-en-2-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents