Please login to your online account to display your discounted pricing

4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  MO01234CB

 View more versions of this product

Catalog No. MO01234CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-(1, 3, 2-Dioxaborinan-2-yl)benzaldehyde
Packaging Amber glass bottle
Assay Percent Range 97%
Molecular Formula C10H11BO3
MDL Number MFCD04039026
CAS 160068-88-2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(1, 3, 2-Dioxaborinan-2-yl)benzaldehyde
CAS 160068-88-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11BO3
Formula Weight 190
MDL Number MFCD04039026
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents