Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(1,2,4-Triazol-4-yl)acetanilide, 99%

$177.01 - $177.01

Chemical Identifiers

Chemical Name or Material 4'-(1,2,4-Triazol-4-yl)acetanilide
CAS 154594-15-7
Molecular Formula C10H10N4O
Formula Weight 202.22
MDL Number MFCD01075653
Beilstein 7639486
Synonym 4-(4-Acetamidophenyl)-1,2,4-triazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1362806

 
alfa aesar™
L1362806
5g Each for $177.01
Description & Specifications

Specifications

Formula Weight 202.22
Chemical Name or Material 4'-(1,2,4-Triazol-4-yl)acetanilide
MDL Number MFCD01075653
CAS 154594-15-7
Synonym 4-(4-Acetamidophenyl)-1,2,4-triazole
Health Hazard 1 Warning
Assay 99%
Molecular Formula C10H10N4O
Beilstein 7639486
Melting Point 284° to 286°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.