missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(1,2,4-Triazol-4-yl)acetanilide, 99%

Manufacturer:  Alfa Aesar™ L1362803

 View more versions of this product

Catalog No. AAL1362803



Specifications

Specifications

4'-(1,2,4-Triazol-4-yl)acetanilide
1g
C10H10N4O
7639486
RBAFMDFJFDEJBG-UHFFFAOYSA-N
N-[4-(1,2,4-triazol-4-yl)phenyl]acetamide
4086562
99%
284°C to 286°C
154594-15-7
MFCD01075653
n-4-1,2,4-triazol-4-yl phenyl acetamide, 4'-1,2,4-triazol-4-yl acetanilide, n-4-4h-1,2,4-triazol-4-yl phenyl acetamide, acetamide, n-4-4h-1,2,4-triazol-4-yl phenyl, n-4-1,2,4-triazol-4-yl phenyl acetamide, n-4-4h-1,2,4-triazol-4-yl phenyl acetamide, 4-1,2,4-triazol-4-yl acetanilide, 4-4-acetamidophenyl-1,2,4-triazole, 4-4-acetylamino phenyl-4h-1,2,4-triazole, acetamide,n-4-4h-1,2,4-triazol-4-yl phenyl
CC(=O)NC1=CC=C(C=C1)N2C=NN=C2
202.217
202.22

Chemical Identifiers

154594-15-7
202.217
RBAFMDFJFDEJBG-UHFFFAOYSA-N
4086562
CC(=O)NC1=CC=C(C=C1)N2C=NN=C2
C10H10N4O
MFCD01075653
n-4-1,2,4-triazol-4-yl phenyl acetamide, 4'-1,2,4-triazol-4-yl acetanilide, n-4-4h-1,2,4-triazol-4-yl phenyl acetamide, acetamide, n-4-4h-1,2,4-triazol-4-yl phenyl, n-4-1,2,4-triazol-4-yl phenyl acetamide, n-4-4h-1,2,4-triazol-4-yl phenyl acetamide, 4-1,2,4-triazol-4-yl acetanilide, 4-4-acetamidophenyl-1,2,4-triazole, 4-4-acetylamino phenyl-4h-1,2,4-triazole, acetamide,n-4-4h-1,2,4-triazol-4-yl phenyl
N-[4-(1,2,4-triazol-4-yl)phenyl]acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents