Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(1,2,4-Triazol-1-yl)phenol, 98%

$34.49 - $432.54

Chemical Identifiers

Chemical Name or Material 4- (1,2,4-Triazol-1-yl)phenol
CAS 68337-15-5
Molecular Formula C8H7N3O
Formula Weight 161.16
MDL Number MFCD00003096
Beilstein 608492
Synonym 1-(4-Hydroxyphenyl)-1,2,4-triazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1210903

 
alfa aesar™
A1210903
1g Each for $34.49

AAA1210906

 
alfa aesar™
A1210906
5g Each for $107.30

AAA1210914

 
alfa aesar™
A1210914
25g Each for $432.54
Description & Specifications

Specifications

Formula Weight 161.16
Molecular Formula C8H7N3O
Melting Point 252° to 257°C
Chemical Name or Material 4- (1,2,4-Triazol-1-yl)phenol
Assay 98%
MDL Number MFCD00003096
CAS 68337-15-5
Beilstein 608492
Synonym 1-(4-Hydroxyphenyl)-1,2,4-triazole
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.