Please login to your online account to display your discounted pricing

4-(1,2,3-Thiadiazol-4-yl)benzamide, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
CAS 175205-53-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3OS
Formula Weight 205.24
MDL Number MFCD00173809
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW01125EA

  SDS
fisher scientific company
SEW01125EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.24
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
CAS 175205-53-5
MDL Number MFCD00173809
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C9H7N3OS