Please login to your online account to display your discounted pricing

4-(1,2,3-Thiadiazol-4-yl)benzamide, 95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW01125EA

 View more versions of this product

Catalog No. SEW01125EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.24
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C9H7N3OS
MDL Number MFCD00173809
CAS 175205-53-5

Specifications

Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
CAS 175205-53-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7N3OS
Formula Weight 205.24
MDL Number MFCD00173809