missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid 98.0+%, TCI America™
Supplier: TCI America T30275G
Specifications
4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid | |
178°C | |
MFCD00155926 | |
SVGWTILJZWYEMD-UHFFFAOYSA-N | |
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid | |
2776639 | |
≥98.0% (GC,T) |
10009-25-3 | |
C9H6F4O3 | |
5 g | |
OC(=O)C1=CC=C(OC(F)(F)C(F)F)C=C1 | |
238.14 | |
238.14 | |
Crystalline Powder |
Chemical Identifiers
10009-25-3 | |
238.14 | |
SVGWTILJZWYEMD-UHFFFAOYSA-N | |
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid |
C9H6F4O3 | |
MFCD00155926 | |
2776639 | |
OC(=O)C1=CC=C(OC(F)(F)C(F)F)C=C1 |
Safety and Handling
TSCA : No