missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid, TCI America™

Manufacturer:  TCI America T302725G

 View more versions of this product

Catalog No. T302725G



Specifications

Specifications

4-(1,1,2,2-Tetrafluoroethoxy)benzoic Acid
178°C
10009-25-3
MFCD00155926
C1=CC(=CC=C1C(=O)O)OC(C(F)F)(F)F
238.138
238.14
≥98.0% (GC,T)
Undesignated
25g
C9H6F4O3
SVGWTILJZWYEMD-UHFFFAOYSA-N
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid
2776639
Crystal-Powder at 20°C

Chemical Identifiers

10009-25-3
238.138
SVGWTILJZWYEMD-UHFFFAOYSA-N
4-(1,1,2,2-tetrafluoroethoxy)benzoic acid
C9H6F4O3
MFCD00155926
2776639
C1=CC(=CC=C1C(=O)O)OC(C(F)F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents