Please login to your online account to display your discounted pricing

(3S,4R)-4-(4'-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (3S, 4R)-4-(4'-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
CAS 105812-81-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13 H18 F N O
Formula Weight 223.29
MDL Number MFCD06658161
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356250050

  SDS
thermo fisher scientific
356250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 223.29
Physical Form Powder
Quantity 5g
Color White
TSCA TSCA
Melting Point 97° to 101°C
Specific Rotation Condition − 38.00 (20.00°C c=1,MeOH)
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Specific Rotation -35° to -41° (20°C, 589nm) (c=1, MeOH)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
MDL Number MFCD06658161
Chemical Name or Material (3S, 4R)-4-(4'-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
CAS 105812-81-5
Molecular Formula C13 H18 F N O
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.