Please login to your online account to display your discounted pricing

Alfa Aesar™ (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol, 98%

Chemical Identifiers

Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol
CAS 105812-81-5
Molecular Formula C13H18FNO
Formula Weight 223.29
MDL Number MFCD06658161
UN Number UN3077
Synonym (3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2813714

 
alfa aesar™
H2813714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2813706

 
alfa aesar™
H2813706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.29
Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol
Assay 98%
Molecular Formula C13H18FNO
MDL Number MFCD06658161
CAS 105812-81-5
UN Number UN3077
Synonym (3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P261-P280b-P262-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves and eye protection.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 93° to 98°C
Quantity 25g
Optical Rotation −34° (c=1 in Methanol)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280b-P262-P273-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves and eye protection.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.