missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3S)-(-)-3-(Methylamino)pyrrolidine, TCI America™

$326.95 - $1,087.80

Chemical Identifiers

CAS 139015-32-0
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00191462
InChI Key NGZYRKGJWYJGRS-YFKPBYRVSA-N
PubChem CID 10261057
IUPAC Name (3S)-N-methylpyrrolidin-3-amine
SMILES CNC1CCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11081G View Documents TCI America
M11081G
1g Each for $326.95
 
M11085G View Documents TCI America
M11085G
5g Each for $1,087.80
 
Specifications

Chemical Identifiers

139015-32-0
100.165
NGZYRKGJWYJGRS-YFKPBYRVSA-N
(3S)-N-methylpyrrolidin-3-amine
C5H12N2
MFCD00191462
10261057
CNC1CCNC1

Specifications

164°C
1g
C5H12N2
2734
CNC1CCNC1
100.165
100.17
≥97.0% (GC,T)
Yellow
139015-32-0
MFCD00191462
NGZYRKGJWYJGRS-YFKPBYRVSA-N
(3S)-N-methylpyrrolidin-3-amine
10261057
Clear Liquid at 20°C
(3S)-(-)-3-(Methylamino)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : No