Please login to your online account to display your discounted pricing

(3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid 97%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  333830050

 View more versions of this product

Catalog No. AC333830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 311.34
Quantity 5g
Color White
TSCA TSCA
Melting Point 136° to 141°C
CAS Max % 100.0
CAS Min % 96.0
Chemical Name or Material (3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00672545
Molecular Formula C18H17NO4
CAS 79261-58-8
Name Note 97%
Assay Percent Range 97%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Name Note 97%
CAS 79261-58-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C18H17NO4
Formula Weight 311.34
MDL Number MFCD00672545
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents