missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid 97%, ACROS Organics™

Chemical Identifiers

CAS 79261-58-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C18H17NO4
Formula Weight 311.34
MDL Number MFCD00672545
Chemical Name or Material (3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC333830050 SDS Thermo Fisher Scientific
333830050
Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

79261-58-8
100.0
C18H17NO4
MFCD00672545
≥96.0%
96.0
97%
311.34
(3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%

Specifications

Crystalline Powder
5g
136° to 141°C
96.0
MFCD00672545
79261-58-8
Authentic
≥96.0%
311.34
White
100.0
Glass Bottle
C18H17NO4
97%
(3S)-2-[Benzyloxycarbonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA