Please login to your online account to display your discounted pricing

Alfa Aesar™ (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one, 98%

$88.76 - $259.16

Chemical Identifiers

Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one
CAS 97538-67-5
Molecular Formula C7H8Cl3NO2
Formula Weight 244.51
MDL Number MFCD08274464
Synonym (2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one; L-Proline-chloral condensation product
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2716403

  SDS
alfa aesar™
H2716403
1g Each for $88.76

AAH2716406

  SDS
alfa aesar™
H2716406
5g Each for $259.16
Description & Specifications

Specifications

Formula Weight 244.51
Melting Point 109°C
Optical Rotation −2.9° (c= 1 in Chloroform)
Molecular Formula C7H8Cl3NO2
Assay 98%
Synonym (2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one; L-Proline-chloral condensation product
Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one
CAS 97538-67-5
Quantity 1g
MDL Number MFCD08274464