Please login to your online account to display your discounted pricing

(3R,6R)-3,6-Octanediol, 95% (98% e.e.), ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  363215000

 View more versions of this product

Catalog No. AC363215000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Needle-Like Crystals
Formula Weight 146.23
Quantity 500mg
Color Off-white
TSCA TSCA
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Synonym (3R, 6R)-Dihydroxyoctane
Chemical Name or Material (3R, 6R)-3, 6-Octanediol
MDL Number MFCD01631087
CAS 129619-37-0
Name Note 95%, (98% e.e.)
Molecular Formula C8H18O2

Specifications

Chemical Name or Material (3R, 6R)-3, 6-Octanediol
Name Note 95%, (98% e.e.)
CAS 129619-37-0
Assay ≥94%
Molecular Formula C8H18O2
Formula Weight 146.23
MDL Number MFCD01631087
Synonym (3R, 6R)-Dihydroxyoctane