missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3R,6R)-3,6-Octanediol, 95% (98% e.e.), ACROS Organics™

Chemical Identifiers

CAS 129619-37-0
Molecular Formula C8H18O2
Formula Weight 146.23
MDL Number MFCD01631087
Synonym (3R, 6R)-Dihydroxyoctane
Chemical Name or Material (3R, 6R)-3, 6-Octanediol, 95%, (98% e.e.)
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC363210010 SDS Thermo Fisher Scientific
363210010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC363215000 SDS Thermo Fisher Scientific
363215000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

129619-37-0
146.23
(3R, 6R)-Dihydroxyoctane
≥94%
C8H18O2
MFCD01631087
(3R, 6R)-3, 6-Octanediol, 95%, (98% e.e.)

Specifications

129619-37-0
Authentic
Needle-Like Crystals
146.23
(3R, 6R)-Dihydroxyoctane
1g
≥94%
97.5% min. (GC)
Glass bottle
C8H18O2
MFCD01631087
Off-white
(3R, 6R)-3, 6-Octanediol, 95%, (98% e.e.)

Safety and Handling

TSCA : TSCA