Please login to your online account to display your discounted pricing

(-)-[3R,4S]-Chromanol 293B, Tocris

IKS blocker; Enantiomer of Chromanol 293B

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material (-)-[3R, 4S]-Chromanol 293B
Assay Percent Range >99%
Molecular Formula C15H20N2O4S
Formula Weight 324.39
Synonym N-[(3R,4S)-6-Cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-methylethanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

147510

 
r & d systems
RDS147510
10mg Each for $246.52

147550

 
r & d systems
RDS147550
50mg Each for $1,023.37
Description & Specifications

Specifications

Formula Weight 324.39
Quantity 10mg
CAS 163163-24-4
Synonym N-[(3R,4S)-6-Cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-methylethanesulfonamide
Molecular Formula C15H20N2O4S
Chemical Name or Material (-)-[3R, 4S]-Chromanol 293B
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature