Please login to your online account to display your discounted pricing

(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%, ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  348722500

 View more versions of this product

Catalog No. AC348722500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder, Crystals, and/or Chunks
Formula Weight 193.24
Quantity 250mg
Color Off-white to yellow to light brown
Melting Point 95° to 101°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Specific Rotation -29° to -35° (20°C, 589nm) (c=1, MeOH)
Assay Percent Range 97%
Chemical Name or Material (3R, 4R)-(-)-1-Benzyl-3, 4-pyrrolidindiol
Molecular Formula C11H15NO2
CAS 163439-82-5
MDL Number MFCD01073894
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (3R, 4R)-(-)-1-Benzyl-3, 4-pyrrolidindiol
CAS 163439-82-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H15NO2
Formula Weight 193.24
MDL Number MFCD01073894
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents