missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3R)-(+)-3-(Methylamino)pyrrolidine, TCI America™

$291.81 - $976.27

Chemical Identifiers

CAS 139015-33-1
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00191461
InChI Key NGZYRKGJWYJGRS-RXMQYKEDSA-N
PubChem CID 10261056
IUPAC Name (3R)-N-methylpyrrolidin-3-amine
SMILES CNC1CCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11071G View Documents TCI America
M11071G
1g Each for $291.81
 
M11075G View Documents TCI America
M11075G
5g Each for $976.27
 
Specifications

Chemical Identifiers

139015-33-1
100.165
NGZYRKGJWYJGRS-RXMQYKEDSA-N
(3R)-N-methylpyrrolidin-3-amine
C5H12N2
MFCD00191461
10261056
CNC1CCNC1

Specifications

164°C
1g
C5H12N2
2734
CNC1CCNC1
100.165
100.17
≥98.0% (GC,T)
Yellow
139015-33-1
MFCD00191461
NGZYRKGJWYJGRS-RXMQYKEDSA-N
(3R)-N-methylpyrrolidin-3-amine
10261056
Clear Liquid at 20°C
(3R)-(+)-3-(Methylamino)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator