Please login to your online account to display your discounted pricing

Alfa Aesar™ (3R)-(+)-3-Acetamidopyrrolidine, 98%

Chemical Identifiers

Chemical Name or Material (3R)-(+)-3-Acetamidopyrrolidine
CAS 131900-62-4
Molecular Formula C6H12N2O
Formula Weight 128.17
MDL Number MFCD00082729
UN Number UN3259
Beilstein 4904138
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1256703

 
alfa aesar™
L1256703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1256706

 
alfa aesar™
L1256706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL12567MD

 
alfa aesar™
L12567MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.17
Chemical Name or Material (3R)-(+)-3-Acetamidopyrrolidine
Assay 98%
Molecular Formula C6H12N2O
MDL Number MFCD00082729
CAS 131900-62-4
Melting Point 52° to 57°C
UN Number UN3259
Beilstein 4904138
Health Hazard 1 Danger
Boiling Point 70° to 72°C (40mmHg)
Quantity 1g
Optical Rotation +46.5° (c=1 in Ethanol)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.