missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3R)-(+)-3-Acetamidopyrrolidine, 98%

Manufacturer:  Alfa Aesar™ L1256706

 View more versions of this product

Catalog No. AAL1256706



Specifications

Specifications

(3R)-(+)-3-Acetamidopyrrolidine
70°C to 72°C (40mmHg)
5g
C6H12N2O
UN3259
3r-+-3-acetamidopyrrolidine, r-n-pyrrolidin-3-yl acetamide, r-3-acetamidopyrrolidine, n-3r-pyrrolidin-3-yl acetamide, acetamide, n-3r-3-pyrrolidinyl, 3r---3-acetamidopyrrolidine, 3r-3-acetamidopyrrolidine, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, r-3-acetamido-pyrrolidine
CC(=O)NC1CCNC1
128.175
128.17
+46.5° (c=1 in Ethanol)
52°C to 57°C
131900-62-4
MFCD00082729
4904138
HDCCJUCOIKLZNM-ZCFIWIBFSA-N
N-[(3R)-pyrrolidin-3-yl]acetamide
7016129
98%

Chemical Identifiers

131900-62-4
128.175
HDCCJUCOIKLZNM-ZCFIWIBFSA-N
7016129
CC(=O)NC1CCNC1
C6H12N2O
MFCD00082729
3r-+-3-acetamidopyrrolidine, r-n-pyrrolidin-3-yl acetamide, r-3-acetamidopyrrolidine, n-3r-pyrrolidin-3-yl acetamide, acetamide, n-3r-3-pyrrolidinyl, 3r---3-acetamidopyrrolidine, 3r-3-acetamidopyrrolidine, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, r-3-acetamido-pyrrolidine
N-[(3R)-pyrrolidin-3-yl]acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

Documents