missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, TCI America™

$142.09 - $427.78

Chemical Identifiers

CAS 147081-49-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD03427030
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-N
Synonym r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
PubChem CID 854070
IUPAC Name tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29201G View Documents TCI America
B29201G
1g Each for $142.09
 
B29205G View Documents TCI America
B29205G
5g Each for $427.78
 
Specifications

Chemical Identifiers

147081-49-0
186.255
CMIBWIAICVBURI-SSDOTTSWSA-N
854070
CC(C)(C)OC(=O)N1CCC(C1)N
C9H18N2O2
MFCD03427030
r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate

Specifications

244°C
1g
C9H18N2O2
r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
CC(C)(C)OC(=O)N1CCC(C1)N
186.255
186.26
≥98.0% (GC,T)
Yellow
147081-49-0
MFCD03427030
CMIBWIAICVBURI-SSDOTTSWSA-N
tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate
854070
Clear Liquid at 20°C
(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
Safety and Handling

Safety and Handling

TSCA : No