3M™ Welding Helmets 10
For Stick and MIG welding processes
$20.11 - $130.21
Specifications
Quantity 1/Cs.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-031-144 3M™
101121
With Auto-Darkening Filter Each for $130.21
19-031-143 3M™
101185
Welding Helmet Without Headband, Auto-Darkening Filter Each for $20.11
Specifications
Description
  • Robust, industrial design with adjustable ratchet headgear
  • Can be worn with select disposable respirators
  • Lithium battery and two user-selectable sensitivity levels
  • Features user-selectable dark shades 10 through 12, with light state of Shade 3
  • Light weight, approximately 13.8 oz. (390g) including auto-darkening welding filter
  • Viewing area: 3.66 x 1.73 in.
  • 1500 hours battery life
Description & Specifications
Specifications
1/Cs.