3M™ Virtua™ Sport Eyewear
Sport styling with maximum protection
Manufacturer:  3M™ 113840000020*
 View more versions of this product
Catalog No. 19-130-3911
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • Polycarbonate lens absorbs 99.9% UV
  • Nine-base curve lens for wrap and front/side coverage
  • Sleek unisex styling in a comfortable, light polycarbonate frame
  • 3M Catalog# 11384-00000-20, 11385-00000-20, 11386-00000-20, 11388-00000-20, 11542-00000-20, 11543-00000-20, 11544-00000-20, 11743-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
ANSI Z87.1-2015, CSA Z94.3-2010
11384-00000-20
Clear
Clear
High-impact Polycarbonate
Anti-fog