missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Versaflo™ Units and Assemblies for TR-300 Series PAPRs

Provide protection against particulates and certain nuisance odors

$630.00 - $1,457.45

Specifications

Color Gray
Filter Type HE Filter
For Use With (Equipment) Tr-300 series powered air purifying respirator
Includes Easy clean belt, high capacity battery, he filter and single-station charger
View More Specs

Includes: Easy clean belt, high capacity battery, HE filter, and single-station charger

Products 3
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
19-039-557
View Documents
3M™
TR305N
PAPR assembly with standard belt and economy battery
Case for $1,307.15
 
19-039-556
View Documents
3M™
TR302N+
PARP assembly without filter, belt, battery, and charger
Case for $630.00
 
19-039-559
View Documents
3M™
TR307N+
PAPR assembly with easy clean belt and high capacity battery
Case for $1,457.45
 
Description

Description

  • The TR-302N PAPR Unit includes only the PAPR blower, filter cover, and flow tube. This allows the customer to choose the Versaflo™ belt, battery, charger, and filter that meets their needs.
  • The TR-305N PAPR assembly contains a TR-302N PAPR Unit, a TR-370 filter cover, a TR-325 belt, a TR-330 battery, a TR-341N battery charger kit, a TR-3710N HE filter, and a TR-3600 Pre-Filter.
  • The TR-306N PAPR assembly contains a TR-302N PAPR Unit, a TR-370 filter cover, a TR-326 belt, a TR-332 battery, a TR-341N battery charger kit, a TR-3710N HE filter, a TR-362 Spark Arrester, and a TR-3600 Pre-Filter.
  • The TR-307N PAPR assembly contains a TR-302N PAPR Unit, a TR-370 filter cover, a TR-327 belt, a TR-332 battery, a TR-341N battery charger kit, a TR-3710N HE filter, and a TR-3600 Pre-Filter.
Specifications

Specifications

Gray
HE Filter
Tr-300 series powered air purifying respirator
Easy clean belt, high capacity battery, he filter and single-station charger
SDS