missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Versaflo TR-300 Series PAPR Headcovers

Easiest way to select a complete system for hospital, healthcare, and pharmaceutical applications where an S-133, APF 25 headcover is preferred

$1,713.10 - $1,713.10

Specifications

Color Gray
Product Type PAPR Kit
For Use With (Equipment) Tr-300 series powered air purifying respirator
Includes Battery, vinyl belt, charger, filter, breathing tube with cover, headcover and carrying/storage bag
Mounting Type Back
View More Specs

Alert:

Before use, the wearer must read and understand the USER Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation. In Canada, CSA standards Z94.4 requirements must be met and/or requirements of the applicable jurisdiction, as appropriate. Misuse may result in sickness or death.


Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-039-554 View Documents 3M™
TR300+HKS
Small/Medium Case for $1,713.10
 
19-039-553 View Documents 3M™
TR300HKL
Medium/Large Case for $1,713.10
 
Description

Description

Kit Components

  • S-133 Headcover
  • TR-307N PAPR with Easy Clean Belt
  • BT-30 Breathing Tube
  • Charger
  • TR-3710N filter
  • Storage bag
  • Sizes: Small/Medium and Medium/Large

Specifications

Specifications

Gray
Tr-300 series powered air purifying respirator
Back
4.3kg
PAPR Kit
Battery, vinyl belt, charger, filter, breathing tube with cover, headcover and carrying/storage bag
9.5 lb.