3M™ Versaflo™ TR-300 PAPR Series Complete System Kit, Heavy Industry
Complete particulate PAPR system for industrial applications where combined respiratory, head, eye and face protection is required or desired
Manufacturer:  3M™ TR300HIK
Catalog No. 19-039-552
Alert:
Before use, the wearer must read and understand the USER Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation. In Canada, CSA standards Z94.4 requirements must be met and/or requirements of the applicable jurisdiction, as appropriate. Misuse may result in sickness or death.
Description
Description
Kit Components
  • M-307 Respiratory Hard Hat†
  • TR-306N PAPR with High Durability Leather Belt (battery charger, TR-3710N filter, and TR-362 spark arrestor/prefilter are included)
  • BT-40 Heavy Duty Breathing Tube
†ANSI Z89.1-2003 Type I Class G
Specifications
Specifications
10 to 12 hr.
Tr-300 series powered air purifying respirator
Hard hat, high durable lather belt, breathing tube, battery charger, filter, spark arrestor/prefilter
1 lb.
Heavy Industry Powered Air Purifying Respirator Kit
Industrial applications
Belt
0.45kg
Documents