3M™ Versaflo™ Air Filtration Panel Accessories: Cartridges
Accessories for 3M Versaflo Air Filtration Panel
$87.97 - $87.97
Specifications
For Use With (Equipment) AP-600 Series Air Filtration Panels
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-031-127 3M™
AP611
Filter Cartridge Each for $87.97
Specifications
Description
For use with 3M Versaflo Air Filtration Panels
Filter Cartridge
For AP-600 Series Air Filtration Panels
CO Catalytic Cartridge
For AP-603 and AP-603PS Air Filtration Panels
Description & Specifications
Specifications
AP-600 Series Air Filtration Panels