3M™ Tri-Flange™ Ear Plugs
Tri-Flange™ flexible flanges adapt to the shape of the ear canal to comfortably reduce exposure to noise. The plastic stem allows the wearer to insert the ear plugs quickly and easily without touching the premolded tips.
Manufacturer:  3M™ 7000052724
 View more versions of this product
Catalog No. 19-130-0470
Description
Description
  • Triple-flanged, premolded ear plugs fit comfortably in a wide range of sizes
  • Soft, flexible flanges conform to the shape of the ear canal
  • No need to roll ear plugs before fitting so tips stay clean even if hands are dirty
  • Washable and reusable
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Yellow
Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
P3001
400 Pairs/Cs
ABS, elastomeric polymer
No
No
Flanged
Class AL
Cloth
Yes
Poly bag
Clear
Corded
Reusable
26 dB
Universal