Please login to your online account to display your discounted pricing

3M™ Transpore Surgical Tape

Offers strong adhesion

$22.28 - $22.28

Specifications

Adhesive Acrylate
Color Clear
Length (English) 10 yd.
Length (Metric) 9m
Material Plastic
View More Specs
Products
Catalog Number Mfr. No. Quantity Width (English) Width (Metric) Price Quantity    

18-999-380

 
3m™
15271/2
24 Pack 0.5 in. 1.25cm Pack of 24 Each for $22.28
Case of 10 Pack for $206.90

18-999-381

 
3m™
15271
12 Pack 1 in. 2.54cm Pack of 12 Each for $22.28
Case of 10 Pack for $206.90
Description & Specifications

Specifications

Adhesive Acrylate
Color Clear
Length (English) 10 yd.
Length (Metric) 9m
Material Plastic
Product Type Surgical Tape
For Use With (Application) For anchoring bulky dressings, securing tubing and devices
Latex Free Yes
Waterproof Yes

  • Easy, straight, bi-directional tear allows clinician to tailor it
  • Porous for breathability
  • Easy to handle with gloves
  • Good adhesive to skin and tubing for secure placement
  • Transparent for easy monitoring
  • Latex-free and hypoallergenic for sensitive patients
  • Water-resistant
  • Plastic