3M™ Supplied-Air Respirator Compatible Breathing Tubes (Loose Fitting Headgear)
Designed for use with 3M™ Supplied Air Hood Systems.
$142.54 - $142.54
Specifications
Type Breathing Tube
View More Specs
Includes: Assembly includes breathing tube, metal thumbscrew clamp, and squeeze clamp.
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
19-021593
View Documents
3M™
00051131070318
Breathing Tube for H-Series hoods
Each for $142.54
Description
Description
For use with the 3M hoods, helmets, headgear and facepieces described in ordering table.
  • Breathing tubes, compressed air supply hoses, and air control devices are required parts of a NIOSH-approved system.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
Breathing Tube