missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Versaflo™ Peel-Off Visor Cover for S-series Hood Assembly
$24.84 - $81.28
Specifications
Color | Clear |
---|---|
For Use With (Application) | Can be applied to certain s series hoods to protect visor from contaminants and scratches |
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity | ||||
19-805-622
|
3M™
7000127434 |
Peel-Off Visor Cover, Medium/Large | Integrated Harness |
Case of 40 Each for $81.28
|
|
19-805-624
|
3M™
7100138630 |
Peel-Off Visor Cover | S-series Hood Assembly |
Pack of 10 Each for $24.84
Case of 4 Pack for $99.36 |
|
Description
- Transparent covering to hood
- Protect from contaminants and scratches
- 3M Catalog# S-920L , 3M Catalog# S-922
Specifications
Clear | |
Can be applied to certain s series hoods to protect visor from contaminants and scratches |