3M™ Versaflo™ Air-Flow Adjustment Cover for Premium Head Suspension
Cover is used to adjust the location of the airflow in the hood by attaching to the end of the breathing tube. For use with S-950 Premium Head Suspension.
Supplier: 3M™ 7100010623
Description
- For use with S-950 Premium Head Suspension
- Attached to end of breathing tube, ring allows user to adjust location of airflow in hood
- 3M Catalog# S-955
Specifications
OSHA | |
Airflow Adjustment Cover | |
S-955 |
Reusable | |
Premium Head Suspension S-950 |