3M™ Ratchet Headgear and Faceshield Safety System
Manufacturer:  3M™ 7000127243
 View more versions of this product
Catalog No. 19-145-465
Description
Description
  • 3M Catalog# 82782-00000 , 3M Catalog# 82783-00000
Specifications
Specifications
Safety System
Safety
Provides impact protection
Propionate
9 x 14.25 in.
Clear
Combination of ratchet headgear and WP96 faceshield; Five position crown strap
Documents