3M™ Qualitative Fit-Test Kit, Sweet
Use to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
$26.85 - $448.48
Specifications
Certifications/Compliance OSHA 29 CFR 1910.134, Appendix A
View More Specs
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Includes: Laminated instruction booklet (multilingual); .9 oz. (55ml) sensitivity solution; 1.9 oz. (55mL) fit test solution; Two nebulizers; Hood; Collar assembly; Two sets replacement nebulizer inserts. Optional training and fit test case includes fit-test kit materials as well as training materials (videos, posters and literature).
Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
17-985-93
View Documents
3M™
7000001898
Qualitative Fit Test Kit 1 Ea.
Each for $448.48
19-039428
View Documents
3M™
7000029587
Sensitivity Solution Refill 6/Cs.
Each for $28.13
Case of 6 Each for $159.00
19-039429
View Documents
3M™
7100004636
Sweet Solution Refill 6/Cs.
Each for $26.85
Case of 6 Each for $151.80
Description
Description
  • Works by spraying a sweet solution into the hood environment
  • If wearer does not detect sweet taste, the fit test has been passed
  • 3M Catalog# FT-10, FT-11, FT-12, FT-20
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Order Info
Individual 1.9 oz. (55mL) bottles of sensitivity solution or fit test solution also available.
Compliance
Meets the performance criteria for fit testing respirators under OSHA 29 CFR 1910.134, Appendix A.
Specifications
Specifications
OSHA 29 CFR 1910.134, Appendix A
SDS