missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Qualitative Fit-Test Kit, Bitter

Contains everything necessary to fit test any particulate respirator or gas/vapor respirator with a particulate prefilter

$22.22 - $352.54

Specifications

Certifications/Compliance OSHA 29 CFR 1910.134, Appendix A
For Use With (Application) Automotive, construction, food and beverage manufacturing, metal production and fabrication, military maintenance, repair and operation, mining, oil and gas, pharmaceutical, transportation
View More Specs

Alert:

Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.


Includes: 1.9 oz. (55mL) sensitivity solution (bitter); 1.9 oz. (55mL) fit test solution (bitter); Testing hood and collar; Two nebulizers; Two sets replacement nebulizer inserts; Detailed, multilingual instruction booklet

Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
19-002722
View Documents
3M™
7100004634
Fit Test Solution 6/Cs.
Each for $22.22
Case of 6 Each for $133.32
 
19-002721
View Documents
3M™
7100004635
Sensitivity Test Solution 6/Cs.
Each for $22.22
Case of 6 Each for $133.32
 
19-002720
View Documents
3M™
7000002042
Bitter Fit Test Kit
Each for $352.54
 
Description

Description

  • Use for fit testing on NIOSH-approved respirators, in accordance with OSHA 29 CFR 1910.134, Appendix A.
  • 3M Catalog# FT-30 , 3M Catalog# FT-31 , 3M Catalog# FT-32

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Order Info

Individual 1.9 oz. (55mL) bottles of sensitivity solution or fit test solution also available.

Specifications

Specifications

OSHA 29 CFR 1910.134, Appendix A
Automotive, construction, food and beverage manufacturing, metal production and fabrication, military maintenance, repair and operation, mining, oil and gas, pharmaceutical, transportation
SDS