missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Pistonz™ Push-in Foam Ear Plugs

NRR 29dB

$336.72 - $336.72

Specifications

Certifications/Compliance CSA Class AL
Color Silver
Product Type Pistonz™ Push-In Foam Earplug
Size One size fits to all
For Use With (Application) Metallic-style earplug tips with ultra-soft feel for all day wear
View More Specs

Alert:

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.


Products 1
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
29-636-3005 View Documents 3M™
P1400
Uncorded Case of 400 Each for $336.72
 
Description

Description

Officially licensed hearing protection of NASCAR™.

  • Instant-use; No need to roll down
  • Stem is stiff enough for insertion, yet designed to flex sideways if impacted
  • Metallic-style earplug tips with ultra-soft feel for all day wear

Specifications

Specifications

CSA Class AL
Pistonz™ Push-In Foam Earplug
Metallic-style earplug tips with ultra-soft feel for all day wear
29dB
Silver
One size fits to all
Polyurethane
Push-to-fit Foam